missing translation for 'onlineSavingsMsg'
Learn More

Bicine, 99.38%, MP Biomedicals™

Product Code. 11338589 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
This item is not returnable. View return policy

Product Code. 11338589

Brand: MP Biomedicals 0210100583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-bis [2-hydroxyethyl]glycine
PubChem CID 8761
ChEBI CHEBI:40957
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 250 g
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
Formula Weight 163.2
Chemical Name or Material Bicine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado