missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BICD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12808742
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808742 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808742 Supplier Invitrogen™ Supplier No. PA520560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is A549 cell lysate. PA5-20560 can be used with blocking peptide PEP-0680.

BICD2 is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BICD2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene BICD2
Gene Accession No. Q8TD16, Q921C5
Gene Alias bA526D8.1; bicaudal D homolog 2; bicaudal D homolog 2 (Drosophila); bicaudal D protein; Bic-D 2; BICD cargo adaptor 2; BICD2; coiled-coil protein BICD2; cytoskeleton-like bicaudal D protein homolog 2; homolog of Drosophila bicaudal D; KIAA0699; Protein bicaudal D homolog 2; RP11-476B13.3; SMALED2
Gene Symbols BICD2
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the center of human BICD2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23299, 306809, 76895
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.