Learn More
Invitrogen™ BICD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587781
Description
Immunogen sequence: HVAVAEAGEL REQLKALRST HEAREAQHAE EKGRYEAEGQ ALTEKVSLLE KASRQDRELL ARLEKELKKV SDVAGETQGS LSVAQDELVT FSEELANLYH HVCMCNNETP NRVMLDYYRE GQGGAGRTSP GGRTSPEARG RRSPILLPKG LLAPEAGRAD GGTGDSSPSP GSSLPSPLSD P; Positive Samples: U-87MG, HL-60, HeLa, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Golgi apparatus, cytoskeleton.
BICD2 is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8.
Specifications
| BICD2 | |
| Polyclonal | |
| Unconjugated | |
| BICD2 | |
| bA526D8.1; bicaudal D homolog 2; bicaudal D homolog 2 (Drosophila); bicaudal D protein; Bic-D 2; BICD cargo adaptor 2; BICD2; coiled-coil protein BICD2; cytoskeleton-like bicaudal D protein homolog 2; homolog of Drosophila bicaudal D; KIAA0699; Protein bicaudal D homolog 2; RP11-476B13.3; SMALED2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23299, 306809, 76895 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.88 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TD16, Q921C5 | |
| BICD2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 440-620 of human BICD2 (NP_0010038001). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.