missing translation for 'onlineSavingsMsg'
Learn More

Bicarbonate Indicator Solution, Pure, Concentrated Stock Reagent, Fisher Chemical™

Product Code. 10234340 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10234340 250 mL 250mL
10486870 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234340 Supplier Fisher Chemical Supplier No. J/2300/07
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Bicarbonate Indicator Solution, Pure, Concentrated Stock Reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 239809

Specifications

Packaging Amber glass bottle
Color Orange to Red
Quantity 250 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.