missing translation for 'onlineSavingsMsg'
Learn More

Bicalutamide, MedChemExpress

Product Code. 16498617
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
500mg
5g
This item is not returnable. View return policy

Product Code. 16498617

Brand: Medchem Express HY14249

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bicalutamide is an orally active non-steroidal androgen receptor (AR) antagonist. Bicalutamide can be used for the research of prostate cancer.

Bicalutamide is an orally active non-steroidal androgen receptor (AR) antagonist. Bicalutamide can be used for the research of prostate cancer.

Specifications

Chemical Name or Material Bicalutamide
CAS 90357-06-5
Color White
Purity Grade Notes Research
Molecular Formula C18H14F4N2O4S
Quantity 5 g
Solubility Information DMSO : ≥ 50 mg/mL (116.18 mM)
SMILES FC1=CC=C(S(=O)(CC(O)(C(NC2=CC=C(C#N)C(C(F)(F)F)=C2)=O)C)=O)C=C1
Molecular Weight (g/mol) 430.37
Formula Weight 430.37
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.