missing translation for 'onlineSavingsMsg'
Learn More

Bicalutamide, MedChemExpress

Product Code. 16471716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
500mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471716 500 mg 500mg
16498617 5 g 5g
16481399 1 g 1g
16471116 200 mg 200mg
16416426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16471716 Supplier Medchem Express Supplier No. HY14249
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bicalutamide is an orally active non-steroidal androgen receptor (AR) antagonist. Bicalutamide can be used for the research of prostate cancer.

Bicalutamide is an orally active non-steroidal androgen receptor (AR) antagonist. Bicalutamide can be used for the research of prostate cancer.

Specifications

Chemical Name or Material Bicalutamide
CAS 90357-06-5
Color White
Purity Grade Notes Research
Molecular Formula C18H14F4N2O4S
Quantity 500 mg
Solubility Information DMSO : ≥ 50 mg/mL (116.18 mM)
SMILES FC1=CC=C(S(=O)(CC(O)(C(NC2=CC=C(C#N)C(C(F)(F)F)=C2)=O)C)=O)C=C1
Molecular Weight (g/mol) 430.37
Formula Weight 430.37
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.