missing translation for 'onlineSavingsMsg'
Learn More

Bicalutamide EP Impurity D, Mikromol™

Product Code. 30049689
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30049689

Brand: Mikromol MM0894.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bicalutamide EP Impurity D
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Amino-2-cyanobenzotrifluoride
CAS 654-70-6
Analyte or Component Names 5-Amino-2-cyanobenzotrifluoride
Shipping Condition Room Temperature
Molecular Formula C8 H5 F3 N2
Certifications/Compliance ISO 17025
Synonym 4-Amino-2-(trifluoromethyl)benzonitrile, Bicalutamide Imp. D (EP), 4-Amino-2-(trifluoromethyl)benzonitrile, 5-Amino-2-cyanobenzotrifluoride, Bicalutamide Impurity D
InChI Formula InChI=1S/C8H5F3N2/c9-8(10,11)7-3-6(13)2-1-5(7)4-12/h1-3H,13H2
SMILES Nc1ccc(C#N)c(c1)C(F)(F)F
IUPAC Name 4-amino-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol) 186.13
Quantity 25 mg
Formula Weight 186.0405 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.