missing translation for 'onlineSavingsMsg'
Learn More

Bicalutamide, 97%, Thermo Scientific Chemicals

Product Code. 15688705 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15688705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15688705 Supplier Thermo Scientific Acros Supplier No. 461000010
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 90357-06-5
Melting Point 196.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 4 h)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C18H14F4N2O4S
Quantity 1 g
Synonym bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex
InChI Key LKJPYSCBVHEWIU-UHFFFAOYSA-N
SMILES CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
Molecular Weight (g/mol) 430.37
PubChem CID 2375
Formula Weight 430.37
Percent Purity 97%
Chemical Name or Material Bicalutamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.