missing translation for 'onlineSavingsMsg'
Learn More

BIBR 1087 SE, MedChemExpress

Product Code. 16472668
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16472668

Brand: Medchem Express HYW004360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BIBR 1087 SE is an intermediate metabolite of dabigatran etexilate.

BIBR 1087 SE is an intermediate metabolite of dabigatran etexilate.

Specifications

Chemical Name or Material BIBR 1087 SE
CAS 212321-78-3
Color White
Purity Grade Notes Research
Molecular Formula C32H37N7O5
Quantity 5 mg
Synonym Desethyl Dabigatran Etexilate
Solubility Information DMSO : ≥ 100 mg/mL (166.76 mM)
SMILES N=C(C1=CC=C(C=C1)NCC2=NC3=CC(C(N(CCC(O)=O)C4=NC=CC=C4)=O)=CC=C3N2C)NC(OCCCCCC)=O
Molecular Weight (g/mol) 599.68
Formula Weight 599.68
Percent Purity 96.57%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.