missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BIBO 3304 trifluoroacetate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BIBO 3304 trifluoroacetate is a high affinity NPY Y1 receptor antagonist (IC50 values are 0.38 and 0.72 nM at human and rat receptors respectively) that displays > 2600-fold selectivity over Y2, Y4 and Y5 receptors. Inhibits NPY - and fasting-induced feeding in vivo following central administration. Also antagonizes anxiolytic-like effects of NPY.
Specifications
Specifications
| Chemical Name or Material | N-[(1R)-1-[[[[4-[[(Aminocarbonyl)amino]methyl]phenyl]methyl]amino]carbonyl]-4-[(aminoiminomethyl)amino]butyl]-α-phenyl-benzeneacetamide ditrifluoroacetate |
| CAS | 2310085-85-7 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C29H35N7O3.2CF3CO2H |
| Formula Weight | Observed MW: 757.69 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction