missing translation for 'onlineSavingsMsg'
Learn More

BIBF 1202, MedChemExpress

Product Code. 16471589
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471589 1 mg 1mg
16419507 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471589 Supplier Medchem Express Supplier No. HY15992
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BIBF 1202 is the carboxylate metabolite of BIBF 1120 which inhibits VEGFR2 kinase with an IC50 of 62 nM.

BIBF 1202 is the carboxylate metabolite of BIBF 1120 which inhibits VEGFR2 kinase with an IC50 of 62 nM.

Specifications

Chemical Name or Material BIBF 1202
CAS 894783-71-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H31N5O4
Quantity 1 mg
Solubility Information DMSO : ≥ 45 mg/mL (85.62 mM)
SMILES O=C(C1=CC(NC/2=O)=C(C=C1)C2=C(NC3=CC=C(N(C)C(CN4CCN(C)CC4)=O)C=C3)/C5=CC=CC=C5)O
Molecular Weight (g/mol) 525.6
Formula Weight 525.6
Percent Purity 96.62%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.