missing translation for 'onlineSavingsMsg'
Learn More

Biapenem, MedChemExpress

Product Code. 16475758
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16475758

Brand: Medchem Express HY1357310mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Biapenem (CLI 86815; L 627; LJC 10627) a parenteral carbapenem antibacterial agent with a broad spectrum.

Biapenem (CLI 86815

Specifications

Chemical Name or Material Biapenem
Physical Form Solid
CAS 120410-24-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H18N4O4S
For Use With (Application) COVID-19-immunoregulation
Synonym CLI 86815 L 627 LJC 10627
Solubility Information H2O : 10 mg/mL (28.54 mM; Need ultrasonic)
SMILES O=C1N2C(C([O-])=O)=C(SC(C3)CN4[N+]3=CN=C4)[C@H](C)[C@]2([H])[C@@]1([H])[C@H](O)C
Molecular Weight (g/mol) 350.39
Formula Weight 350.39
Percent Purity 98.31%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.