missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies BI-D1870

Product Code. 17150112
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17150112

Brand: STEMCELL Technologies 72714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

90 kDa ribosomal S6 kinase (RSK) inhibitor

  • BI-D1870 inhibits the 90 kDa ribosomal S6 kinases (RSKs), which are serine/threonine kinases involved in diverse cellular processes including growth, survival, and motility (Romeo et al.)
  • Reduces neural stem cell proliferation and self-renewal in vitro (Karelina et al.)
  • Inhibits growth of breast cancer cell lines (Stratford et al.), Dhillon et al.)

Specifications

CAS 501437-28-1
Quantity 10 mg
Molecular Formula C19H23F2N5O2
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.