missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies BI-D1870
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- BI-D1870 inhibits the 90 kDa ribosomal S6 kinases (RSKs), which are serine/threonine kinases involved in diverse cellular processes including growth, survival, and motility (Romeo et al.)
- Reduces neural stem cell proliferation and self-renewal in vitro (Karelina et al.)
- Inhibits growth of breast cancer cell lines (Stratford et al.), Dhillon et al.)
Specifications
Specifications
| CAS | 501437-28-1 |
| Quantity | 1 mg |
| Molecular Formula | C19H23F2N5O2 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction