missing translation for 'onlineSavingsMsg'
Learn More

(±)-BI-D, MedChemExpress

Product Code. 16497476
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497476 100 mg 100mg
16463547 50 mg 50mg
16460008 5 mg 5mg
16447528 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497476 Supplier Medchem Express Supplier No. HY18601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ((plusmn))-BI-D is a potent ALLINI(An allosteric IN inhibitor) that binds integrase at the LEDGF/p75 binding site.

((plusmn))-BI-D is a potent ALLINI(An allosteric IN inhibitor) that binds integrase at the LEDGF/p75 binding site.

Specifications

Chemical Name or Material (±)-BI-D
CAS 1416258-16-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H27NO4
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (246.62 mM)
SMILES OC(C(OC(C)(C)C)C1=C(C(C=CC=C2)=C2N=C1C)C3=CC(CCCO4)=C4C=C3)=O
Molecular Weight (g/mol) 405.49
Formula Weight 405.49
Percent Purity 95.66%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.