missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BI-749327, MedChemExpress
Description
BI-749327 is a potent, high selectivity and orally bioavailable TRPC6 antagonist, with IC50s of 13 nM, 19 nM and 15 nM for mouse, human and guinea pig TRPC6, respectively. BI-749327 is 85-fold more selective for mouse TRPC6 than TRPC3 and 42-fold versus TRPC7.
Specifications
Specifications
| Chemical Name or Material | BI-749327 |
| CAS | 2361241-23-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H21F3N4O2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 31.25 mg/mL (70.63 mM; Need ultrasonic) |
| SMILES | NC1=NN=C(C2CCN(C(C3=CC=C(OC4=CC=C(C(F)(F)F)C=C4)C=C3)=O)CC2)C=C1 |
| Molecular Weight (g/mol) | 442.43 |
| Formula Weight | 442.43 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?