missing translation for 'onlineSavingsMsg'
Learn More

BI 689648, MedChemExpress

Product Code. 16446027
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16446027

Brand: Medchem Express HY101217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BI 689648 is a novel, highly selective aldosterone synthase inhibitor which can inhibit CYP11B1 and CYP11B2 with IC50s of 310 and 2.1 nM, respectively.

BI 689648 is a novel, highly selective aldosterone synthase inhibitor which can inhibit CYP11B1 and CYP11B2 with IC50s of 310 and 2.1 nM, respectively.

Specifications

Chemical Name or Material BI 689648
CAS 1633009-87-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H18N4O2
Quantity 5 mg
Solubility Information DMSO : 30 mg/mL (100.56 mM; Need ultrasonic and warming)
SMILES O=C(N1CCCC2=C1N=CC(C3=CC(COC)=CN=C3)=C2)N
Molecular Weight (g/mol) 298.34
Formula Weight 298.34
Percent Purity 98.39%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.