missing translation for 'onlineSavingsMsg'
Learn More

BI-671800, MedChemExpress

Product Code. p-200096771
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485586

Brand: Medchem Express HY114141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BI-671800 is a highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2), with IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively. BI-671800 has potential for the treatment of poorly controlled asthma.

BI-671800 is a highly specific and potent antagonist of chemoattractant receptor-homologous molecule on Th2 cells (DP2/CRTH2), with IC50 values of 4.5 nM and 3.7 nM for PGD2 binding to CRTH2 in hCRTH2 and mCRTH2 transfected cells, respectively. BI-671800 has potential for the treatment of poorly controlled asthma.

Specifications

Chemical Name or Material BI-671800
Physical Form Solid
CAS 1093108-50-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H26F3N5O3
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 135 mg/mL (269.19 mM; Need ultrasonic)
SMILES O=C(O)CC1=C(N(C)C)N=C(CC2=CC=C(NC(C3=CC=C(C(F)(F)F)C=C3)=O)C=C2)N=C1N(C)C
Molecular Weight (g/mol) 501.5
Formula Weight 501.5
Percent Purity 99.23%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.