missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BI-3802, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
BI-3802 is a highly potent BCL6 degrader and inhibits the Bric-à-brac (BTB) domain of BCL6 with an IC50 of ≤3 nM. BI-3802 induces the polymerization of BCL6 and promotes BCL6 degration depended on E3 ligase SIAH1. BI-3802 has antitumor activity.
Specifications
Specifications
| Chemical Name or Material | BI-3802 |
| Physical Form | Solid |
| CAS | 2166387-65-9 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H29ClN6O3 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 5 mg/mL (10.31 mM; Need ultrasonic) |
| SMILES | ClC1=CN=C(N2C[C@H](C)C[C@H](C)C2)N=C1NC3=CC(C=C(OCC(NC)=O)C(N4C)=O)=C4C=C3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction