missing translation for 'onlineSavingsMsg'
Learn More

BI-3802, MedChemExpress

Product Code. 16476947
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476947

Brand: Medchem Express HY108705

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BI-3802 is a highly potent BCL6 degrader and inhibits the Bric-à-brac (BTB) domain of BCL6 with an IC50 of ≤3 nM. BI-3802 induces the polymerization of BCL6 and promotes BCL6 degration depended on E3 ligase SIAH1. BI-3802 has antitumor activity.

BI-3802 is a highly potent BCL6 degrader and inhibits the Bric-à-brac (BTB) domain of BCL6 with an IC50 of ≤3 nM. BI-3802 induces the polymerization of BCL6 and promotes BCL6 degration depended on E3 ligase SIAH1. BI-3802 has antitumor activity.

Specifications

Chemical Name or Material BI-3802
Physical Form Solid
CAS 2166387-65-9
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H29ClN6O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 5 mg/mL (10.31 mM; Need ultrasonic)
SMILES ClC1=CN=C(N2C[C@H](C)C[C@H](C)C2)N=C1NC3=CC(C=C(OCC(NC)=O)C(N4C)=O)=C4C=C3
Molecular Weight (g/mol) 484.98
Formula Weight 484.98
Percent Purity 99.43%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.