missing translation for 'onlineSavingsMsg'
Learn More

BI-1347, MedChemExpress

Product Code. 16477677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477677 5 mg 5mg
16454938 10 mg 10mg
16435716 100 mg 100mg
16421187 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477677 Supplier Medchem Express Supplier No. HY120350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BI-1347 is a potent CDK8 inhibitor extracted from patent WO2017202719A1, product I-003, has an IC50 of 1.1 nM.

BI-1347 is a potent CDK8 inhibitor extracted from patent WO2017202719A1, product I-003, has an IC50 of 1.1 nM.

Specifications

Chemical Name or Material BI-1347
CAS 2163056-91-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H20N4O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (350.71 mM; Need ultrasonic)
SMILES O=C(N(C)C)CN1N=CC(C2=CC=C(C3=CN=CC4=C3C=CC=C4)C=C2)=C1
Molecular Weight (g/mol) 356.42
Formula Weight 356.42
Percent Purity 98.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.