missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bi-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582588
This item is not returnable.
View return policy
Description
Immunogen sequence: MNIFDRKINF DALLKFSHIT PSTQQHLKK Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
BI-1 (Bax Inhibitor 1) is in anti-apoptotic protein that has been linked to protection from apoptosis induced endoplasmic reticulum (ER) stress (reviewed in Bailley-Maitre et al, 2007). BI-1 contains several transmembrane domains, localizes to ER membranes, and has cytoprotective functions that are conserved in both animals and plants. BI-1 suppresses apoptosis induced by ectopic expression of the proapoptotic protein Bax as well as other types of stimuli. Cells from BI-1 knockout (KO) mice are hypersensitivite to apoptosis induced by ER stress-causing chemical agents (thapsigargin, tunicamycin, and brefeldin A) or by ischemia-reperfusion (IR) injury. Conversely, overexpression of BI-1 protects against apoptosis induced by ER stress and IR. The mechanism by which BI-1 protects cells from ER-stress induced apoptosis remains to be fully elucidated, itis thought to involve regulation of Ca2+ handling by the ER.
Specifications
| Bi-1 | |
| Polyclonal | |
| Unconjugated | |
| TMBIM6 | |
| 5031406P05Rik; Ab1-011; Ac1-149; Bax inhibitor 1; Bax inhibitor-1; BAXI1; Bi1; BI-1; Cc1-27; Tegt; testis enhanced gene transcript; testis-enhanced gene transcript protein; Tmbim6; transmembrane BAX inhibitor motif containing 6; transmembrane BAX inhibitor motif-containing protein 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7009 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55061 | |
| TMBIM6 | |
| Recombinant protein corresponding to Human TMBIM6. Recombinant protein control fragment (Product #RP-91281). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction