missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bi-1 Monoclonal Antibody (20F565)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516225
This item is not returnable.
View return policy
Description
Bi-1 Monoclonal Antibody for Western Blot, IHC (P)
BI-1 (Bax Inhibitor 1) is in anti-apoptotic protein that has been linked to protection from apoptosis induced endoplasmic reticulum (ER) stress (reviewed in Bailley-Maitre et al, 2007). BI-1 contains several transmembrane domains, localizes to ER membranes, and has cytoprotective functions that are conserved in both animals and plants. BI-1 suppresses apoptosis induced by ectopic expression of the proapoptotic protein Bax as well as other types of stimuli. Cells from BI-1 knockout (KO) mice are hypersensitivite to apoptosis induced by ER stress-causing chemical agents (thapsigargin, tunicamycin, and brefeldin A) or by ischemia-reperfusion (IR) injury. Conversely, overexpression of BI-1 protects against apoptosis induced by ER stress and IR. The mechanism by which BI-1 protects cells from ER-stress induced apoptosis remains to be fully elucidated, itis thought to involve regulation of Ca2+ handling by the ER.
Specifications
| Bi-1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| P55061, P55062, Q9D2C7 | |
| TMBIM6 | |
| A synthetic peptide made to the N-terminal portion of the human Bax inhibitor 1 protein (between amino acids 1-40). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| 20F565 | |
| Unconjugated | |
| TMBIM6 | |
| 5031406P05Rik; Ab1-011; Ac1-149; Bax inhibitor 1; Bax inhibitor-1; BAXI1; Bi1; BI-1; Cc1-27; Tegt; testis enhanced gene transcript; testis-enhanced gene transcript protein; Tmbim6; transmembrane BAX inhibitor motif containing 6; transmembrane BAX inhibitor motif-containing protein 6 | |
| Mouse | |
| Protein G | |
| RUO | |
| 110213, 24822, 7009 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction