missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BI 01383298

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
BI 01383298 is a potent and selective inhibitor of human SLC13A5 (Na+/citrate cotransporter; IC50 = 56 nM in HEK cells overexpressing hSLC13A5 and 24 nM in HepG2 cells with endogenous SLC13A5 expression). Displays >1000-fold selectivity over other family members.
Specifications
Specifications
| Chemical Name or Material | 1-[(3,5-Dichlorophenyl)sulfonyl]-N-[(4-fluorophenyl)methyl]-4-piperidinecarboxamide |
| CAS | 2227549-00-8 |
| Quantity | 25 mg |
| Molecular Formula | C19H19Cl2FN2O3S |
| Formula Weight | 445.34 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction