missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BI 01383298
GREENER_CHOICE

Product Code. 18787436
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18787436

Brand: Tocris Bioscience™ 6518/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective SLC13A5 (Na+/citrate cotransporter) inhibitor

BI 01383298 is a potent and selective inhibitor of human SLC13A5 (Na+/citrate cotransporter; IC50 = 56 nM in HEK cells overexpressing hSLC13A5 and 24 nM in HepG2 cells with endogenous SLC13A5 expression). Displays >1000-fold selectivity over other family members.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[(3,5-Dichlorophenyl)sulfonyl]-N-[(4-fluorophenyl)methyl]-4-piperidinecarboxamide
CAS 2227549-00-8
Quantity 25 mg
Molecular Formula C19H19Cl2FN2O3S
Formula Weight 445.34
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.