missing translation for 'onlineSavingsMsg'
Learn More

BI 01383298, MedChemExpress

Product Code. p-200096586
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467917 5 mg 5mg
16445886 100 mg 100mg
16415198 10 mg 10mg
16413316 25 mg 25mg
16411417 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16467917 Supplier Medchem Express Supplier No. HY124738

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BI 01383298 is a potent inhibitor of the sodium-citrate co-transporter (SLC13A5) that is highly expressed in the liver.

BI 01383298 is a potent inhibitor of the sodium-citrate co-transporter (SLC13A5) that is highly expressed in the liver.

Specifications

Chemical Name or Material BI 01383298
CAS 2227549-00-8
Color White
Purity Grade Notes Research
Molecular Formula C19H19Cl2FN2O3S
Quantity 5 mg
Solubility Information DMSO : 37.5 mg/mL (84.21 mM; Need ultrasonic)
SMILES O=C(C1CCN(S(=O)(C2=CC(Cl)=CC(Cl)=C2)=O)CC1)NCC3=CC=C(F)C=C3
Molecular Weight (g/mol) 445.34
Formula Weight 445.34
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.