missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BHPI

Description
BHPI is an estrogen receptor (ER) α inhibitor. Inhibits proliferation of drug-resistant ERα-positive breast and ovarian cancer cells. Induces sustained activation of the unfolded protein response. Active in vivo.
Specifications
Specifications
| Chemical Name or Material | 1,3-Dihydro-3,3-bis(4-hydroxyphenyl)-7-methyl-2H-indol-2-one |
| CAS | 56632-39-4 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Molecular Formula | C21H17NO3 |
| Formula Weight | Observed MW: 331.36 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?