missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHPI, MedChemExpress
Description
BHPI is a potent inhibitor of nuclear estrogen-ERα-regulated gene expression
Specifications
Specifications
| Chemical Name or Material | BHPI |
| CAS | 56632-39-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H17NO3 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 120 mg/mL (362.14 mM; Need ultrasonic) |
| SMILES | O=C1NC2=C(C=CC=C2C)C1(C3=CC=C(O)C=C3)C4=CC=C(O)C=C4 |
| Molecular Weight (g/mol) | 331.36 |
| Formula Weight | 331.36 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?