missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BHMT2 Polyclonal specifically detects BHMT2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | BHMT2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | betaine-homocysteine methyltransferase 2, betaine--homocysteine S-methyltransferase 2, EC 2.1.1.5, FLJ20001 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human BHMT2 (NP_001171476.1). Peptide sequence FTFSASEDNMESKWEDVNAAACDLAREVAGKGDALVAGGICQTSIYKYQK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?