missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHMT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15965-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocystinemia, but such a defect has not yet been observed.Specifications
| BHMT | |
| Polyclonal | |
| Unconjugated | |
| BHMT | |
| BHMT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12116, 635 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35490, Q93088 | |
| Bhmt | |
| BHMT Fusion Protein Ag8738 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction