missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BHMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114904
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BHMT.
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.
Specifications
| BHMT | |
| Polyclonal | |
| Unconjugated | |
| BHMT | |
| betaine-homocysteine methyltransferase; betaine--homocysteine S-methyltransferase; betaine-homocysteine S-methyltransferase; betaine--homocysteine S-methyltransferase 1; BHMT; BHMT1; epididymis secretory sperm binding protein Li 61p; HEL-S-61p | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12116, 635, 81508 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O09171, O35490, Q93088 | |
| BHMT | |
| A synthesized peptide derived from human BHMT(Accession Q93088), corresponding to amino acid residues D371-Q406. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction