missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BHMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521466
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, mouse liver. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (86%), Pig (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.
Specifications
| BHMT | |
| Polyclonal | |
| Unconjugated | |
| BHMT | |
| betaine-homocysteine methyltransferase; betaine--homocysteine S-methyltransferase; betaine-homocysteine S-methyltransferase; betaine--homocysteine S-methyltransferase 1; BHMT; BHMT1; epididymis secretory sperm binding protein Li 61p; HEL-S-61p | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12116, 635 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O35490, Q93088 | |
| BHMT | |
| Recombinant protein encompassing a sequence within the center region of human BHMT. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction