missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BHMT Monoclonal Antibody (OTI3E11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525054
This item is not returnable.
View return policy
Description
BHMT Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.
Specifications
| BHMT | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q93088 | |
| BHMT | |
| Full length human recombinant protein of BHMT produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI3E11 | |
| Unconjugated | |
| BHMT | |
| betaine-homocysteine methyltransferase; betaine--homocysteine S-methyltransferase; betaine-homocysteine S-methyltransferase; betaine--homocysteine S-methyltransferase 1; BHMT; BHMT1; epididymis secretory sperm binding protein Li 61p; HEL-S-61p | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 635 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction