missing translation for 'onlineSavingsMsg'
Learn More

BHMT Antibody (OTI3E11), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18433613 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433613 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433613 Supplier Novus Biologicals Supplier No. NBP270250AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BHMT Monoclonal antibody specifically detects BHMT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHMT
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3E11
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias betaine homocysteine methyltransferase, betaine-homocysteine methyltransferase, betaine--homocysteine S-methyltransferase, betaine--homocysteine S-methyltransferase 1, BHMT1, EC 2.1.1, EC 2.1.1.5
Host Species Mouse
Immunogen Full length recombinant protein of human BHMT (NP_001704) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 635
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.