missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHMT Antibody (3D6), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04264R
This item is not returnable.
View return policy
Description
BHMT Monoclonal specifically detects BHMT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| BHMT | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BHMT | |
| Recombinant human BHMT (1-406aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| NP_001704 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 3D6 | |
| Western Blot, ELISA | |
| betaine homocysteine methyltransferase, betaine-homocysteine methyltransferase, betaine--homocysteine S-methyltransferase, betaine--homocysteine S-methyltransferase 1, BHMT1, EC 2.1.1, EC 2.1.1.5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 635 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction