missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRA13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597288
This item is not returnable.
View return policy
Description
Immunogen sequence: MEGAGAGSGF RKELVSRLLH LHFKDDKTKE AAVRGVRQAQ AEDALRVDVD QLEKVLPQLL LDF; Positive Samples: H460; Cellular Location: Chromosome, Nucleus, centromere, kinetochore.
STRA13 is a DNA-binding component of the FA core complex involved in DNA damage repair and genome maintenance.STRA13 is recruited to forks stalled by DNA interstrand cross-links, and required for cellular resistance to such lesions. As a component of the APITD1/CENPS complex, STRA13 is also essential for the stable assembly of the outer kinetchore.
Specifications
| STRA13 | |
| Polyclonal | |
| Unconjugated | |
| CENPX | |
| CENPX; CENP-X; Centromere protein X; D9; FAAP10; FANCM-associated histone fold protein 2; FANCM-interacting histone fold protein 2; Fanconi anemia-associated polypeptide of 10 kDa; MGC14480; MHF2; retinoic acid-inducible gene D9 protein homolog; stimulated by retinoic acid 13; stimulated by retinoic acid 13 homolog; stimulated by retinoic acid gene 13 protein homolog; STRA13 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 201254 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A8MT69 | |
| CENPX | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction