missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BHLHB5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564152
This item is not returnable.
View return policy
Description
Immunogen sequence: AAAAALHPAL GAYEQAAGYP FSAGLPPAAS CPEKCALFNS VSSSLCKQCT E Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
BHLHB5 inhibits DNA binding of TCF3/E47 homodimers and TCF3 (E47)/NEUROD1 heterodimers and acts as a strong repressor of Neurod1 and Myod-responsive genes, probably by heterodimerization with class a basic helix-loop-helix factors. BHLHB5 despite the presence of an intact basic domain, does not bind to DNA.
Specifications
| BHLHB5 | |
| Polyclonal | |
| Unconjugated | |
| BHLHE22 | |
| basic helix-loop-helix (bHLH) gene, class B, beta3; basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix domain containing, class B5; basic helix-loop-helix family member e22; basic helix-loop-helix family, member e22; Beta3; BHLHB5; bHLHe22; CAGL85; Class B basic helix-loop-helix protein 5; class E basic helix-loop-helix protein 22; Protein BETA3; TNRC20; trinucleotide repeat containing 20; trinucleotide repeat-containing gene 20 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFJ8 | |
| BHLHE22 | |
| Recombinant protein corresponding to Human BHLHB5. Recombinant protein control fragment (Product #RP-103544). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction