missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BHLHA9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564042
This item is not returnable.
View return policy
Description
Immunogen sequence: SKARRMAANV RERKRILDYN EAFNALRRAL RHDLGGKRLS KIATLRRAIH RIAALSLVLR ASPAPRGPCG HLECHG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Putative transcription factor, which may play a role in limb development.
Specifications
| BHLHA9 | |
| Polyclonal | |
| Unconjugated | |
| BHLHA9 | |
| A830053O21Rik; basic helix-loop-helix family member a9; basic helix-loop-helix family, member a9; bHLHa9; bHLHf42; class A basic helix-loop-helix protein 9; Class B basic helix-loop-helix factor 42; class F basic helix-loop-helix factor 42; class II basic helix-loop-helix protein; Fingerin; novel helix-loop-helix DNA-binding domain containing protein; Orphan1a; wu:fb99e06 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 727857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7RTU4 | |
| BHLHA9 | |
| Recombinant protein corresponding to Human BHLHA9. Recombinant protein control fragment (Product #RP-103460). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction