missing translation for 'onlineSavingsMsg'
Learn More

Melford BHI Broth

Product Code. p-200087062
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 30018059

Brand: Melford B110001000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recommended for the cultivation of rare microorganisms.

Recommended for the cultivation of a wide variety of microorganisms including many types of pathogens. BHI Agar is currently recommended as a general medium for aerobic bacteria and for the primary recovery of fungi from clinical specimens.

Dissolve 50 grams in 1L distilled water. Sterilize by autoclaving 15 minutes.

Ingredients:

Brain Heart Infusion from solids: 17.5 g/l

Pancreatic Digest of Gelatin: 10.0 g/l

Dextrose: 2.0 g/l

Sodium Chloride: 5.0 g/l

Disodium Phosphate: 2.5 g/l

Agar: 13.5 g/l

Specifications

Quantity 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.