missing translation for 'onlineSavingsMsg'
Learn More

BG45, MedChemExpress

Product Code. 16477598
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477598 10 mg 10mg
16463617 50 mg 50mg
16410078 5 mg 5mg
16407526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477598 Supplier Medchem Express Supplier No. HY18712
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BG45 is an HDAC class I inhibitor with selectivity for HDAC3 (IC50 = 289 nM).

BG45 is an HDAC class I inhibitor with selectivity for HDAC3 (IC50 = 289 nM).

Specifications

Chemical Name or Material BG45
CAS 926259-99-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H10N4O
Quantity 10 mg
Solubility Information DMSO : ≥ 48 mg/mL (224.07 mM)
SMILES O=C(C1=NC=CN=C1)NC2=CC=CC=C2N
Molecular Weight (g/mol) 214.22
Formula Weight 214.22
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.