missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BFSP2 Polyclonal Antibody

Description
Immunogen sequence: LSRNYEEDVK LLHKQLAGCE LEQMDAPIGT GLDDILETIR IQWERDVEKN RVEAGALLQA KQQAEVAHMS QTQEEKLAAA LRVE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, the protein product of this gene (phakinin), and filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene have been associated with juvenile-onset, progressive cataracts and Dowling-Meara epidermolysis bullosa simplex.
Specifications
Specifications
| Antigen | BFSP2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BFSP2 |
| Gene Accession No. | Q13515 |
| Gene Alias | 49 kDa cytoskeletal protein; AI448593; beaded filament protein CP49; beaded filament structural protein 2; beaded filament structural protein 2, phakinin; bfps2, Cytoskeletal protein, 49-kD; BFSP2; CP 47; CP47; CP49; CTRCT12; DKEYP-73D8.2; hypothetical protein LOC494090; lens fiber cell beaded filament protein CP 47; Lens fiber cell beaded filament protein CP 49; lens intermediate filament-like light; LIFL-L; MGC142078; MGC142080; phakinin; PHAKOSIN; zgc:103750 |
| Gene Symbols | BFSP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?