missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BFSP2 Polyclonal Antibody

BFSP2 Polyclonal Antibody
Brand: Invitrogen PA588949
This item is not returnable.
View return policy
Description
BFSP2 Polyclonal Antibody for Western Blot
Specifications
| BFSP2 | |
| Polyclonal | |
| Unconjugated | |
| BFSP2 | |
| 49 kDa cytoskeletal protein; AI448593; beaded filament protein CP49; beaded filament structural protein 2; beaded filament structural protein 2, phakinin; bfps2, Cytoskeletal protein, 49-kD; BFSP2; CP 47; CP47; CP49; CTRCT12; DKEYP-73D8.2; hypothetical protein LOC494090; lens fiber cell beaded filament protein CP 47; Lens fiber cell beaded filament protein CP 49; lens intermediate filament-like light; LIFL-L; MGC142078; MGC142080; phakinin; PHAKOSIN; zgc:103750 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 107993 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.11 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6NVD9 | |
| BFSP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 116-415 of human BFSP2 (NP_0035621). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction