missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Made of high purity cotton linters for uniform buffer flow and resulting blots
- No additives to avoid interference during transfer
- Supplied in sheets
Specifications
Specifications
| Length (Metric) | 580 mm |
| Width (Metric) | 600 mm |
| For Use With (Application) | Transfer DNA or RNA according to the Southern technique or semidry blotting of proteins |
| Weight | 550 g/m2 |
| Color | White |
| Quantity | 25/Pk. |
| Material | Cotton linters and refined pulp with an alpha-cellulose content |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?