Learn More
Sartorius™ BF4 Cotton Linter Blotting Paper
High purity cotton linters offer the maximum degree of absorptiveness and cellulose content. With no additives they avoid any interference during the transfer, making them the ideal complement to nitrocellulose blotting membranes.
Brand: Sartorius™ FT-2-521-580580G
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Made of high purity cotton linters for uniform buffer flow and resulting blots
- No additives to avoid interference during transfer
- Supplied in sheets
Specifications
580 mm | |
Transfer DNA or RNA according to the Southern technique or semidry blotting of proteins | |
White | |
Cotton linters and refined pulp with an alpha-cellulose content |
580 mm | |
550 g/m2 | |
25/Pk. | |
1.3 mm |