missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ BF3 Cotton Linter Blotting Paper
Shop All Sartorius Products
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
Description
- Made of high purity cotton linters for uniform buffer flow and resulting blots
- No additives to avoid interference during transfer
- Supplied in sheets
Specifications
Specifications
| Length (Metric) | 460 mm |
| Width (Metric) | 570 mm |
| For Use With (Application) | Increasing and maintaining the transport of liquid from the buffer and as a buffer reservoir in capillary and semidry blotting methods |
| Weight | 330 g/m3 |
| Color | White |
| Quantity | 50/Pk. |
| Format | Sheet |
| Material | Cotton linters with an alpha-cellulose content |
| Thickness | 0.76 mm |
| Grade | BF 3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction