missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BF 2649 hydrochloride
GREENER_CHOICE

Product Code. 18095382
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18095382

Brand: Tocris Bioscience™ 3743/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

H3 receptor inverse agonist/antagonist

Potent and selective histamine H3 receptor inverse agonist (EC50 = 1.5 nM, Ki = 0.16 nM). Exhibits nootropic effects in cognitive disorders; reduces locomotor hyperactivity induced by methamphetamine. Brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 903576-44-3
Molecular Formula C17H27Cl2NO

Specifications

Quantity 50 mg
Formula Weight 332.31
Purity >99%
Molecular Weight (g/mol) 332.309
Chemical Name or Material BF 2649 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.