missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BF 2649 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective histamine H3 receptor inverse agonist (EC50 = 1.5 nM, Ki = 0.16 nM). Exhibits nootropic effects in cognitive disorders; reduces locomotor hyperactivity induced by methamphetamine. Brain penetrant.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 332.31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 332.309 |
| Chemical Name or Material | BF 2649 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction