missing translation for 'onlineSavingsMsg'
Learn More

Bexarotene, MedChemExpress

Product Code. 16451716
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451716 500 mg 500mg
16436406 100 mg 100mg
16412127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451716 Supplier Medchem Express Supplier No. HY14171
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bexarotene (LGD1069) is a high-affinity and selective retinoid X receptors (RXR) agonist with EC50s of 33, 24, 25 nM for RXRα, RXRβ, and RXRγ, respectively. Bexarotene shows limited affinity for RAR receptors (EC50 >10000 nM). Bexarotene can be used for the research of cutaneous T-cell lymphoma.

Bexarotene (LGD1069) is a high-affinity and selective retinoid X receptors (RXR) agonist with EC50s of 33, 24, 25 nM for RXRα, RXRβ, and RXRγ, respectively. Bexarotene shows limited affinity for RAR receptors (EC50 >10000 nM). Bexarotene can be used for the research of cutaneous T-cell lymphoma.

Specifications

Chemical Name or Material Bexarotene
CAS 153559-49-0
Color White
Purity Grade Notes Research
Molecular Formula C24H28O2
Quantity 500 mg
Synonym LGD1069
Solubility Information DMSO : 60 mg/mL (172.18 mM; Need ultrasonic)
SMILES CC1(C2=C(C(C)(CC1)C)C=C(C(C(C3=CC=C(C=C3)C(O)=O)=C)=C2)C)C
Molecular Weight (g/mol) 348.48
Formula Weight 348.48
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.