missing translation for 'onlineSavingsMsg'
Learn More

Bexarotene, 98%, Thermo Scientific Chemicals

Product Code. 15485879
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485879 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15485879 Supplier Thermo Scientific Alfa Aesar Supplier No. J63701.MC
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic retinoid analog with specific affinity for the retinoid X receptor

Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer's model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer′s model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.

Solubility
Soluble in DMSO and ethanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153559-49-0
Molecular Formula C24H28O2
Molecular Weight (g/mol) 348.49
MDL Number MFCD00932428
InChI Key NAVMQTYZDKMPEU-UHFFFAOYSA-N
Synonym bexarotene, targretin, targrexin, targretyn, targret, bexarotenum, bexarotene usan, bexaroteno, bexaroten, targretin-gel
PubChem CID 82146
ChEBI CHEBI:50859
IUPAC Name 4-[1-(3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl]benzoic acid
SMILES CC1=CC2=C(C=C1C(=C)C1=CC=C(C=C1)C(O)=O)C(C)(C)CCC2(C)C

Specifications

Melting Point 220°C to 225°C
Color White
Quantity 100 mg
Merck Index 14,1194
Solubility Information Soluble in DMSO and ethanol
Formula Weight 348.48
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Bexarotene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.