Learn More
Invitrogen™ BEX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139964
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RFRVRQPILQ YRWDIMHRLG EPQARMREEN MERIGEEVRQ LME Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 77%.
This gene belongs to the brain expressed X-linked gene family. The encoded protein interacts with the transcription factor LIM domain only 2 in a DNA-binding complex that recognizes the E-box element and promotes transcription. This gene has been found to be a tumor suppressor that is silenced in human glioma. In breast cancer cells, this gene product modulates apoptosis in response to estrogen and tamoxifen, and enhances the anti-proliferative effect of tamoxifen. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| BEX2 | |
| Polyclonal | |
| Unconjugated | |
| BEX2 | |
| BEX1; BEX2; brain expressed X-linked 2; brain-expressed X-linked protein 2; DJ79P11.1; hBex2; protein BEX2; RP1-79P11.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84707 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXY8 | |
| BEX2 | |
| Recombinant protein corresponding to Human BEX2. Recombinant protein control fragment (Product #RP-99265). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.